Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Natriumhydroxid NF, Multi-Compendial, J.T.Baker®

Natriumhydroxid NF, Multi-Compendial, J.T.Baker®

500 g
Katalog # 3718-32
Ætsende
Leverandør:   Avantor
Natriumhydroxid NF, Multi-Compendial, J.T.Baker®
Katalog # 3718-32
Leverandør:  Avantor
CAS nummer:  1310-73-2
Natriumhydroxid NF, Multi-Compendial, J.T.Baker®

500 g

CAS nummer:  1310-73-2
Price
821,00 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents
Synonymer:  Caustic soda
  • CAS nummer:1310-73-2
  • Formel:NaOH
  • MDL nummer:MFCD00003548
  • Molecular Weight:40 g/mol
  • Merck Index:14,08627
  • Kogende Pt:1390 °C (1013 hPa)
  • Smeltende Pt:323 °C
  • Tæthed:2,13 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:215-185-5
  • FN:1823
  • ADR:8,II
  • NÅ:01-2119457892-27
Specifikationer

Specifikationstestresultater

  • Appearance of solution:
    passes test
  • Assay:
    97.0 - 100.5 %
  • Carbonates:
    max. 2.0 %
  • Chlorides:
    max. 200 ppm
  • Heavy Metals:
    max. 20 ppm
  • Identification A:
    passes test
  • Identification B:
    passes test
  • Iron:
    max. 10 ppm
  • Sulphates:
    max. 200 ppm
  • Chloride:
    max. 0.050 %
  • Clarity and Color of Solution:
    passes test
  • Identification 1:
    passes test
  • Identification 2:
    passes test
  • Mercury:
    passes test
  • Potassium:
    passes test
  • Sodiumcarbonate:
    max. 2.0 %
  • Identification:
    passes test
  • Insoluble substances and organic matters:
    passes test

Specifikationer

  • Pakn:
    500 g

Specifikationer