Cryo.s Cryogenic Storage Tubes with Datamatrix
Product Details & Documents
Manufactured from high quality polypropylene, Cryo.s freezing tubes are suitable for storage of cell cultures, tissue samples, microbiological samples, material of human or animal origin such as blood, serum, and sperm, as well as antibodies, RNA, DNA, and protein samples.
- Sterile
- Endotoxin-free
- Free of RNase, DNase, and human DNA
- Non-pyrogenic and non-cytotoxic
These cryogenic sample tubes are manufactured from high quality polypropylene, which means they are ideally suited for use over a broad range of temperatures (–196 to 121 °C).
Laser etched, ECC200 code is the newest version of Datamatrix coding that is available on Greiner Bio One tubes. Datamatrix coding supports advanced error correction algorithms with capability to decipher partially damaged codes.
Compatible with automated liquid handling and de-/re-capping systems.
: Cryo.s tubes are not intended for sample storage within reproductive medicine.
Documents
Certifikater
Søg efter partinummerSpecifikationer
- Kasket inkluderet:Yes
- Diameter×H:12,5×48 mm
- Kapacitet:2,00 ml
- Beskrivelse:Cryo.s tube with internal thread and silicone gasket
- Lågfarve:Colour-21390331|Colour-21390353
- Grundlag:Round bottom, starfoot base
- Cat. No.:479-2440
- Miljøcertificering:ISO 14001 (241002); ISO 50001 (241002)
- Environmentally Preferable:Lav fremstillingspåvirkning
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



