VWR®, IOS W-Plan Phase Contrast Objectives
IOS W-Plan phase contrast objective, PH 20×/0,40, 5,1 mm
Katalog # 630-1977
Leverandør: VWR Collection
Gå til:
Product Details & Documents
IOS W-Plan PH objectives deliver outstanding performance in phase contrast technique, providing a great contrast generally required in high-level education and routine laboratory needs.
The objectives are designed to ensure field flatness up to F.N. 22, based on an infinity-corrected optical system.
Documents
VWR® for Microscopy
Produktlitteratur
Specifikationer
- Forstørrelse:PH 20×/0,40
- Beskrivelse:IOS W-Plan phase contrast objective
- Seq:208
- Arbejdsafstand:5,1 mm