Orders LinkContactUsLinkComponent
VWR®, IOS W-Plan Phase Contrast Objectives
VWR®, IOS W-Plan Phase Contrast Objectives
IOS W-Plan phase contrast objective, PH 20×/0,40, 5,1 mm
Katalog # 630-1977
Leverandør:   VWR Collection
VWR®, IOS W-Plan Phase Contrast Objectives
Katalog # 630-1977
Leverandør:  VWR Collection
VWR®, IOS W-Plan Phase Contrast Objectives
VWR®, IOS W-Plan Phase Contrast Objectives
IOS W-Plan phase contrast objective, PH 20×/0,40, 5,1 mm
Price
1.930,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
IOS W-Plan PH objectives deliver outstanding performance in phase contrast technique, providing a great contrast generally required in high-level education and routine laboratory needs.
The objectives are designed to ensure field flatness up to F.N. 22, based on an infinity-corrected optical system.
Documents
Specifikationer
  • Forstørrelse:
    PH 20×/0,40
  • Beskrivelse:
    IOS W-Plan phase contrast objective
  • Seq:
    208
  • Arbejdsafstand:
    5,1 mm