Tubes in polystyrene with screw cap
19 ml, 150 mm, 16 mm, on racks sterile packed, Sterile
Leverandør: KIMA SAS DI CHIARIN RENZO
Gå til:
Product Details & Documents
Polystyrene, clear, with red screw cap.
- Sterile, individually or in the rack with 108 tubes in sterile packed available
- Non graduated
- Good thermal and chemical resistance
Universal tubes with round bottom for a wide range of applications.
Documents
Specifikationer
- Udv.Ø:16 mm
- Pakket:on racks sterile packed
- Model:Sterile
- Kapacitet:19 ml
- Længde:150 mm