Orders LinkContactUsLinkComponent
Dunke, Biotainer®, Nalgene®
Dunke, Biotainer®, Nalgene®
5 l
Katalog # NALG3405-99
Leverandør:   Thermo Fisher Scientific
Dunke, Biotainer®, Nalgene®
Katalog # NALG3405-99
Dunke, Biotainer®, Nalgene®
Dunke, Biotainer®, Nalgene®
5 l
Price
Spørg for pris
Log ind for tilgængelighed
1 * 6 Items
Product Details & Documents
PC, clear, with white screw cap.
  • Silicone-lined polypropylene screw cap
  • Good chemical resistance
  • Good temperature resistance
Reduce the risk of carry-over contamination and eliminate the cost of cleaning, sterilizing and associated validations. Use sterile, ready-to-use Biotainer™ Carboys, designed specifically for pharmaceutical and biological manufacturers. Excellent for freezing applications.
Specifikationer
  • Kapacitet:
    5 l