M Stand for Bio-bin® Boxes
M stand for 2 and 5 l Bio-bin® boxes, stainless steel, for benchtop use
Katalog # 119-0468
Leverandør: ECONIX LTD
Gå til:
Product Details & Documents
These stainless steel stands were designed to allow the 2, 5 and 30 Litre Bio-bin® boxes to tilt at approximately 15 degrees to allow easy filling for tall thin items e.g. pipettes.
- Allow the user to see the contents easily while helping to prevent overfilling and also 'wigwamming' of items
- Small stand for 2 and 5 L boxes is ideal for use on work benches
- Larger 30 L M stand is perfect for floor use
Specifikationer
- Beskrivelse:M stand for 2 and 5 l Bio-bin® boxes, stainless steel, for benchtop use