Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
EPA 524.3 reference standards

EPA 524.3 reference standards

524.3 Internal standard/surrogate mix, 2000 µg/ml, P&T Methanol, 1 ml/ampoule, 1 mL
Katalog # REST30017
Leverandør:   Restek
EPA 524.3 reference standards
Katalog # REST30017
Leverandør:  Restek
CAS nummer:  
EPA 524.3 reference standards
EPA 524.3 reference standards

524.3 Internal standard/surrogate mix, 2000 µg/ml, P&T Methanol, 1 ml/ampoule, 1 mL

CAS nummer:  
Price
817,00 kr
Log ind for tilgængelighed
1 * 1 mL
Product Details & Documents

Full 82-component EPA 524.3 list using as few as three ampules, reduce prep time and chances for error or contamination.

  • EPA 524.3 VOA MegaMix ampul includes oxygenates group, no need to order separately
  • Volatile gases prepared separately, replace shorter life components without wasting money on full list
  • Two options for internal and surrogate standards, separate or combined mix
  • Certified reference materials (CRMs) manufactured and QC-tested in Restek’s ISO-accredited labs—satisfy your ISO requirements
  • Also ideal for surface water and groundwater testing

In addition, this collection of certified reference materials (CRMs) also covers the seven volatile organic compounds (VOCs) included in the Unregulated Contaminant Monitoring Rule 3 (UCMR3), which requires monitoring of all public drinking water systems with 10000 or more customers.

Specifikationer

Specifikationer

  • Koncentration:
    2000 µg/ml
  • Matrice:
    P&T Methanol
  • Pakn:
    1 mL
  • Beskrivelse:
    524.3 Internal standard/surrogate mix
  • Pk Info:
    1 ml/ampoule

Specifikationer