Test tubes, flat bottom, without rim, heavy wall
40 mm, 10 mm
Katalog # HECH92277901450
Leverandør: GLASWARENFABRIK KARL HECHT
Test tubes, flat bottom, without rim, heavy wall
Katalog # HECH92277901450
Leverandør: GLASWARENFABRIK KARL HECHT
Gå til:
Product Details & Documents
Made of AR-Glas® (soda glass).
- Heavy wall
- Flat bottom
- Good chemical and physical properties
Documents
HECH42779070_TDS_EN
Datablad
Specifikationer
- Udv.Ø:10 mm
- Længde:40 mm