Orders LinkContactUsLinkComponent
Test tubes, flat bottom, without rim, heavy wall
Test tubes, flat bottom, without rim, heavy wall
40 mm, 10 mm
Katalog # HECH92277901450
Test tubes, flat bottom, without rim, heavy wall
Katalog # HECH92277901450
Test tubes, flat bottom, without rim, heavy wall
Test tubes, flat bottom, without rim, heavy wall
40 mm, 10 mm
Price
640,00 kr
Log ind for tilgængelighed
1 * 500 Items
Product Details & Documents
Made of AR-Glas® (soda glass).
  • Heavy wall
  • Flat bottom
  • Good chemical and physical properties
Documents
Specifikationer
  • Udv.Ø:
    10 mm
  • Længde:
    40 mm