Wafer casettes
4 inch wafer cassette, Rustfrit stål
Katalog # PALBPCPC0101
Leverandør: Palbam
Gå til:
Product Details & Documents
The palbam class stainless steel wafer cassette is designed to replace standard plastic cassettes, in the semiconductor manufacturing process. The cassette has been designed with H Bar.
- Non-contaminating
- Non-deforming
- Fully conductive
- Withstands temperatures up to 400 ºC
- Designed for both manual and robotic handling
Documents
Palbam Class - Catalog 2018
Produktlitteratur
Specifikationer
- Beskrivelse:4 inch wafer cassette
- Materiale:Rustfrit stål
- Seq:01