Galtek® check valves
Inlet 1/4" FNPT, outlet 1/4" Quikgrip®, length 62,7 mm
Catalog # ENTECK4-4N-4Q
Supplier: ENTEGRIS
Jump to:
Product Details & Documents
O-ring free design, ¹/₂ or ¹/₄" orifice.
- O-ring free design reduces contamination
- All wetted parts are molded PFA and PTFE for corrosion resistance
- No metal parts are used that could corrode or contribute to contamination
- Poppet design allows for a high flow through rate and maximises rinsability
Specifications
- Beskrivelse:Inlet 1/4" FNPT, outlet 1/4" Quikgrip®, length 62,7 mm