Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Spray gun

Recirculating tee fitting (9,5 mm Flaretek® tube fitting × 6,4 mm MNPT × 3,2 mm compression)
Katalog # ENTE215-128
Leverandør:   ENTEGRIS
Spray gun
Katalog # ENTE215-128
Leverandør:  ENTEGRIS
Spray gun

Recirculating tee fitting (9,5 mm Flaretek® tube fitting × 6,4 mm MNPT × 3,2 mm compression)

Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

High purity PFA (all wetted parts).

  • With 4,8 mm orifice and ³/₈" Flaretek® tube fitting or ¹/₂" compression connections
  • Max. pressure 5,52 bar

Used in fume hoods and wet processing stations to help protect against corrosion.

Specifikationer
  • Beskrivelse:
    Recirculating tee fitting (9,5 mm Flaretek® tube fitting × 6,4 mm MNPT × 3,2 mm compression)
  • Seq:
    05
Accessories

Produktinformation

  • Katalog #
    Tilgængelighed
    Price
  • Repair tool to access front and rear diaphragm of gun
    Repair tool to access front and rear diaphragm of gun
    ENTE213-80
    Log ind for tilgængelighed
    1 * 1 Items

Produktinformation

  • Katalog #
    Tilgængelighed
    Price
  • Repair kit, includes piston, pin-trigger, front and rear diaphragms, spring, spool
    Repair kit, includes piston, pin-trigger, front and rear diaphragms, spring, spool
    ENTE421-06
    Log ind for tilgængelighed
    1 * 1 Items

Produktinformation

  • Katalog #
    Tilgængelighed
    Price
  • Replacement cap, natural
    Replacement cap, natural
    ENTE600037
    Log ind for tilgængelighed
    1 * 1 Items

Produktinformation

  • Katalog #
    Tilgængelighed
    Price
  • Replacement nozzle
    Replacement nozzle
    ENTE500121
    Log ind for tilgængelighed
    1 * 1 Items

Produktinformation

  • Katalog #
    Tilgængelighed
    Price
  • Replacement cap, orange
    Replacement cap, orange
    ENTE600037OR
    Log ind for tilgængelighed
    1 * 1 Items

Produktinformation

  • Katalog #
    Tilgængelighed
    Price
  • Replacement cap, blue
    Replacement cap, blue
    ENTE600037BL
    Log ind for tilgængelighed
    1 * 1 Items