Desiccator cabinets
Product Details & Documents
Bench top N₂ purge desiccator cabinets made from stainless steel 304, units have an electropolished finish and static dissipative PVC window. Cabinets cut nitrogen expenses by up to 80%, automate clean, dry benchtop storage eliminating moisture & oxygen related degradation and optimise yields. An appropriate flow of nitrogen is introduced into the upper section of the desiccator; the heavier air is then purged out of the bottom. Ideal for use with semiconductor components, biological and pharmaceutical samples, and other sensitive materials. Fully integrated turnkey system takes 30 s to install.
- Built in RH% sensor and display panel
- Easy to use control panel allows user to enter the required RH% level
- Can be used as a single unit on bench top or shelf or as units of two or three in dedicated shelf units
- N₂ desiccator cabinets minimise the N₂ usage and therefore minimise N₂ leakage into the cleanroom atmosphere
Delivery information: Typically available from stock for direct shipment from the supplier.
Documents
Palbam Class - Catalog 2018
Product Literature
Specifikationer
- B×D×H udv.:600×400×400 mm
- Beskrivelse:Bench top N₂ purge desiccator cabinet, with static dissipative PVC window
- Seq:01
- med:RH% sensor and display panel
Accessories
Produktinformation
- Katalog #TilTilgængelighedPrice
- Shelf, solid stainless steel, 800×500 mmShelf, solid stainless steel, 800×500 mmPALBDS8050Desiccator UCD8050Log ind for tilgængelighed1 * 1 Items
Produktinformation
- Katalog #TilTilgængelighedPrice
- Shelf, solid stainless steel, 600×400 mmShelf, solid stainless steel, 600×400 mmPALBDS-6040Desiccator UCD6040Log ind for tilgængelighed1 * 1 Items
Produktinformation
- Katalog #TilTilgængelighedPrice
- Desiccator stand, stainless steel, for three unitsDesiccator stand, stainless steel, for three unitsPALBDSTAND604040-3Desiccator UCD604040Log ind for tilgængelighed1 * 1 Items
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



