Squeegee mop and handles
Telescopical handle, length 2,2 - 4,0 m
Katalog # PPSA3590210
Leverandør: PFENNIG REINIGUNGSTECHNIK
Gå til:
Product Details & Documents
Squeegee mops and handles for hygienically sensitive areas.
- Mop: PP (body), TPE (lip)
- Hygienic squeegee is suitable with cleanroom and telescopical handles
- Handles are anodised aluminum tubes
Specifikationer
- Beskrivelse:Telescopical handle, length 2,2 - 4,0 m
- Seq:03