Stopcocks, Galtek®
Inlet 1/4" MNPT, outlet 1/4" MNPT, length 43,2 mm
Catalog # ENTEPV4-4-2
Supplier: ENTEGRIS
Jump to:
Product Details & Documents
Two- or three way, ¹/₈" orifice.
- All wetted parts are PFA and PTFE.
- Small compact design provides minimal internal volume.
- Several connection sizes available to choose from
Specifications
- Beskrivelse:Inlet 1/4" MNPT, outlet 1/4" MNPT, length 43,2 mm