Nitrogen spray gun
Nitrogen spray gun, orange
Katalog # ENTE421-42-31
Leverandør: ENTEGRIS
Gå til:
Product Details & Documents
Long lasting and reliable PFA nitrogen spray guns with 0,5 µm PTFE membrane for use in high purity and corrosive chemical applications.
- Integral filter enables air to be filtered before nitrogen is dispensed
- Vented cap/nozzle meets OSHA safety specifications
- PFA construction enables use in high purity and corrosive applications
- Colored caps enable visual chemical differentiation at a glance
Nitrogen cap/nozzle prevents pressure build-up if outlet is blocked.
Orifice 4,8 mm
Max. pressure 75 psig (517 kPa)
Connection ¹/₄" FNPT
: Repair kits are available.
Specifikationer
- Beskrivelse:Nitrogen spray gun, orange
- Seq:03
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Repair tool to access front and rear diaphragm of gunRepair tool to access front and rear diaphragm of gunENTE213-80Log ind for tilgængelighed1 * 1 Items
Produktinformation
- Katalog #TilgængelighedPrice
- Replacement cap/nozzle, blue
Produktinformation
- Katalog #TilgængelighedPrice
- Replacement cap/nozzle, natural
Produktinformation
- Katalog #TilgængelighedPrice
- Replacement cap/nozzle, orange