Forceps
Curved, tapered thin tip, 127 mm
Katalog # ENTEC14-0315
Leverandør: ENTEGRIS
Gå til:
Product Details & Documents
ETFE, white. Durable forceps for substrate handling.
- Different tip styles available
- Packaged in protective cases
Specifikationer
- Model:Curved, tapered thin tip
- Længde:127 mm
- Seq:04