Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Spray gun

Spray gun

PFA tubing, coiled, O.Ø 9,5 mm (stretches to 8 ft. long and non flared tubing)
Katalog # ENTE221-60
Leverandør:   ENTEGRIS
Spray gun
Katalog # ENTE221-60
Leverandør:  ENTEGRIS
Spray gun

PFA tubing, coiled, O.Ø 9,5 mm (stretches to 8 ft. long and non flared tubing)

Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

High purity PFA (all wetted parts).

  • With 4,8 mm orifice and ³/₈" Flaretek® tube fitting or ¹/₂" compression connections
  • Max. pressure 5,52 bar

Used in fume hoods and wet processing stations to help protect against corrosion.

Specifikationer
  • Beskrivelse:
    PFA tubing, coiled, O.Ø 9,5 mm (stretches to 8 ft. long and non flared tubing)
  • Seq:
    09
Accessories

Produktinformation

  • Katalog #
    Tilgængelighed
    Price
  • Repair tool to access front and rear diaphragm of gun
    Repair tool to access front and rear diaphragm of gun
    ENTE213-80
    Log ind for tilgængelighed
    1 * 1 Items

Produktinformation

  • Katalog #
    Tilgængelighed
    Price
  • Repair kit, includes piston, pin-trigger, front and rear diaphragms, spring, spool
    Repair kit, includes piston, pin-trigger, front and rear diaphragms, spring, spool
    ENTE421-06
    Log ind for tilgængelighed
    1 * 1 Items

Produktinformation

  • Katalog #
    Tilgængelighed
    Price
  • Replacement cap, natural
    Replacement cap, natural
    ENTE600037
    Log ind for tilgængelighed
    1 * 1 Items

Produktinformation

  • Katalog #
    Tilgængelighed
    Price
  • Replacement nozzle
    Replacement nozzle
    ENTE500121
    Log ind for tilgængelighed
    1 * 1 Items

Produktinformation

  • Katalog #
    Tilgængelighed
    Price
  • Replacement cap, orange
    Replacement cap, orange
    ENTE600037OR
    Log ind for tilgængelighed
    1 * 1 Items

Produktinformation

  • Katalog #
    Tilgængelighed
    Price
  • Replacement cap, blue
    Replacement cap, blue
    ENTE600037BL
    Log ind for tilgængelighed
    1 * 1 Items

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.