Orders LinkContactUsLinkComponent
Galtek® check valves
Inlet 1/2" Flaretek®, outlet 1/2" Flaretek®, length 116,8 mm
Katalog # ENTECK8-8F-8F
Leverandør:   ENTEGRIS
Galtek® check valves
Katalog # ENTECK8-8F-8F
Leverandør:  ENTEGRIS
Galtek® check valves
Inlet 1/2" Flaretek®, outlet 1/2" Flaretek®, length 116,8 mm
Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
O-ring free design, ¹/₂ or ¹/₄" orifice.
  • O-ring free design reduces contamination
  • All wetted parts are molded PFA and PTFE for corrosion resistance
  • No metal parts are used that could corrode or contribute to contamination
  • Poppet design allows for a high flow through rate and maximises rinsability
Specifikationer
  • Beskrivelse:
    Inlet 1/2" Flaretek®, outlet 1/2" Flaretek®, length 116,8 mm