Galtek® check valves
Inlet 1/2" FNPT, outlet 1/2" FNPT, length 84,3 mm
Katalog # ENTECK8-8N-8N
Leverandør: ENTEGRIS
Gå til:
Product Details & Documents
O-ring free design, ¹/₂ or ¹/₄" orifice.
- O-ring free design reduces contamination
- All wetted parts are molded PFA and PTFE for corrosion resistance
- No metal parts are used that could corrode or contribute to contamination
- Poppet design allows for a high flow through rate and maximises rinsability
Specifikationer
- Beskrivelse:Inlet 1/2" FNPT, outlet 1/2" FNPT, length 84,3 mm