Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Cleanroom tapes, Supertack™

Cleanroom tape, hvid, 152,4 mm
Katalog # UTAP1153WH600-P3D
Leverandør:   ULTRATAPE
Cleanroom tapes, Supertack™
Katalog # UTAP1153WH600-P3D
Leverandør:  ULTRATAPE
Cleanroom tapes, Supertack™
Cleanroom tapes, Supertack™

Cleanroom tape, hvid, 152,4 mm

Price
757,00 kr
Log ind for tilgængelighed
1 * 1 ROL
Product Details & Documents

These Supertack™ cleanroom tapes are made of PE with rubber adhesive and are ideal for any application that requires a strong bond.

  • Tapes remove cleanly with no residue
  • Tear by hand
  • Accept cleanroom ballpoint pen writing

The tape is available in Protocol™ uniquely striped color pattern which allows immediate recognition of the product as a certified cleanroom tape. Suitable for use in ISO Class 4 environments

Længde: 32,9 m.

Forskellige farver tilgængelige.

 : Leveres i poser a to stk.

Specifikationer
  • Bredde:
    152,4 mm
  • Farve:
    hvid
  • Beskrivelse:
    Cleanroom tape
  • Seq:
    27