Spring boxes
Holds ten trays and one cover or eleven trays, Polypropylene, natural
Katalog # ENTEH20-SB11-0615
Leverandør: ENTEGRIS
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:Holds ten trays and one cover or eleven trays, Polypropylene, natural
- Cat. No.:ENTEH20-SB11-0615