Orders LinkContactUsLinkComponent
Galtek® check valves
Galtek® check valves
Inlet 3/8" Flaretek®, outlet 3/84" Flaretek®, length 89,7 mm"
Katalog # ENTECK4-6F-6F
Leverandør:   ENTEGRIS
Galtek® check valves
Katalog # ENTECK4-6F-6F
Leverandør:  ENTEGRIS
Galtek® check valves
Inlet 3/8" Flaretek®, outlet 3/84" Flaretek®, length 89,7 mm"
Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
O-ring free design, ¹/₂ or ¹/₄" orifice.
  • O-ring free design reduces contamination
  • All wetted parts are molded PFA and PTFE for corrosion resistance
  • No metal parts are used that could corrode or contribute to contamination
  • Poppet design allows for a high flow through rate and maximises rinsability
Specifikationer
  • Beskrivelse:
    Inlet 3/8" Flaretek®, outlet 3/84" Flaretek®, length 89,7 mm"