Wipes, pre-saturated
203×152 mm, 30 foil packages/box
Katalog # EPTA6-C86-EFP
Leverandør: ESSENTRA POROUS TECHNOLOGIES
Gå til:
Product Details & Documents
These heavyweight polyester/cellulose wipes are pre-saturated with 96% IPA and 4% DIW. They are ideal for general and manufacturing applications, as well as for industrial cleaning.
- Pre-wetted with high concentration of alcohol
- Individually packed: Easy tear, single use foil package
- Dissolves most residues safely and effectively
- Excellent as a degreaser
- Made in the USA
: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904
Specifikationer
- Pakket:30 foil packages/box
- L×B:203×152 mm
- Seq:02