Laboratory coats, PP
With elastic wrist, without pockets, hvid, XL
Katalog # 114-9017
Leverandør: HUBEI ORIENT INTL TRADING
Gå til:
Product Details & Documents
These disposable laboratory coats with shirt neck made from PP and are ideal for non critical applications.
- Elasticated or Velcro wrist
- Press stud fastening (four/five snaps)
Weight: 25 to 55 g/m²
Specifikationer
- Tøj størrelse:XL
- Farve:hvid
- Beskrivelse:With elastic wrist, without pockets
- Seq:10
- Køn:Unisex
- Cat. No.:114-9017
- Vægt:25 - 55 g/m²
- Outfit applikationer:Laboratory
- Lukkestørrelse:Med trykknapper
- Lommer:uden lommer