Shoe covers, CPE
Shoe covers, 9 g/piece, blå, 5 pieces/roll, 10 rolls/poly bag, 10 bags/carton
Katalog # 114-9037
Leverandør: HUBEI ORIENT INTL TRADING
Gå til:
Product Details & Documents
These disposable shoe covers are made of soft and durable CPE.
- CPE is water resistant and provides good resistance against chemicals
- Lightweight, breathable, dust- and waterproof
- Elasticated at the ankle
- Different packaging options available
Suitable for use in laboratories, hospitals, food processing, manufacturing and construction areas.
Specifikationer
- Pakket:5 pieces/roll, 10 rolls/poly bag, 10 bags/carton
- Farve:blå
- Beskrivelse:Shoe covers, 9 g/piece
- Seq:05
- Cat. No.:114-9037
- Outfit applikationer:Laboratory|Food