Orders LinkContactUsLinkComponent
Accessories for centrifuges
Adapter for 4×2 small blood bag/cell culture bags (<350 ml), for TX-1000 rotor, 250 ml conical bottle or 200 ml Thermo Scientific™ Nunc™ Bottle or 175 ml Thermo Scientific™ Nalgene™ conical bottle, Ø×L: 62×145 mm
Katalog # 521-3418
Leverandør:   Thermo Scientific
Accessories for centrifuges
Katalog # 521-3418
Leverandør:  Thermo Scientific
Accessories for centrifuges
Accessories for centrifuges
Adapter for 4×2 small blood bag/cell culture bags (<350 ml), for TX-1000 rotor, 250 ml conical bottle or 200 ml Thermo Scientific™ Nunc™ Bottle or 175 ml Thermo Scientific™ Nalgene™ conical bottle, Ø×L: 62×145 mm
Price
5.420,00 kr
Log ind for tilgængelighed
1 * 4 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Adapter for 4×2 small blood bag/cell culture bags (<350 ml), for TX-1000 rotor, 250 ml conical bottle or 200 ml Thermo Scientific™ Nunc™ Bottle or 175 ml Thermo Scientific™ Nalgene™ conical bottle, Ø×L: 62×145 mm
  • Seq:
    03