Accessories for centrifuges
Adapter for 4×2 small blood bag/cell culture bags (<350 ml), for TX-1000 rotor, 250 ml conical bottle or 200 ml Thermo Scientific™ Nunc™ Bottle or 175 ml Thermo Scientific™ Nalgene™ conical bottle, Ø×L: 62×145 mm
Katalog # 521-3418
Leverandør: Thermo Scientific
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:Adapter for 4×2 small blood bag/cell culture bags (<350 ml), for TX-1000 rotor, 250 ml conical bottle or 200 ml Thermo Scientific™ Nunc™ Bottle or 175 ml Thermo Scientific™ Nalgene™ conical bottle, Ø×L: 62×145 mm
- Seq:03