Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Wall-mounted cabinets

Wall-mounted cabinet, with one shelf, 1 door, right hinged, 450×366×780 mm
Katalog # KOTT307-00047
Leverandør:   KOETTERMANN
Wall-mounted cabinets
Katalog # KOTT307-00047
Leverandør:  KOETTERMANN
Wall-mounted cabinets
Wall-mounted cabinets

Wall-mounted cabinet, with one shelf, 1 door, right hinged, 450×366×780 mm

Price
2.890,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Made of epoxy powder coated steel, with one shelf.

  • Solid hinged doors or sliding glass doors
  • Height adjustable shelfs
  • Maximum loading capacity: 80 kg
Specifikationer
  • Beskrivelse:
    Wall-mounted cabinet, with one shelf
  • Seq:
    17
  • B×D×H:
    450×366×780 mm
  • Døre:
    1 door, right hinged
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.