Accessories
Bottle neck adapter, threaded PP, O.Ø 38 mm, for 525/ 530 models
Katalog # SOCO1.525.GL38
Leverandør:   Socorex Isba
Accessories
Katalog # SOCO1.525.GL38
Leverandør:  Socorex Isba
Accessories
Accessories
Bottle neck adapter, threaded PP, O.Ø 38 mm, for 525/ 530 models
Price
133,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Bottle neck adapter, threaded PP, O.Ø 38 mm, for 525/ 530 models
  • Seq:
    09