Stoppers for tubes
Grip stopper, 16 mm O.D., ufarvet
Catalog # GREI310321BE
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
GRIP CAP 16MM 1 * 100 Items Catalog #212-0642
Product 212-0642 is a replacement for GREI310321BE and has the same packaging and specifications, but may be a different quantity.
Product Details & Documents
Specifications
- Til rør:16 mm O.D.
- Farve:ufarvet
- Beskrivelse:Grip stopper
- Seq:06