PE stoppers, DURAN®
orange, 34/45, 54 mm
Katalog # 214-0402
Leverandør: DWK Life Sciences
Gå til:
Product Details & Documents
These PE stoppers have an ergonomic grip that makes the opening and closing of the flasks easier and more secure. Coloured inserts in the different sizes of stoppers allow the easy identification of the corresponding ground (NS/TS) neck size.
: DIN 12254
Specifikationer
- NS:34/45
- Farve:orange
- Højde:54 mm
- Seq:08