Insole, Action Dry
Insoles, Action Dry, 35, sort
UPOWCS00015/35
: U-Power
Made from EVA/textile.
- Light non-slip
- Keeps feet dry
- Anti-fungal treated
- Skostørrelse:35
- Farve:sort
- Beskrivelse:Insoles, Action Dry
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
#: 20864.320
: VWR Chemicals
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
#: 83640.320
: VWR Chemicals
Natriumhydroxid 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM Reag. Ph. Eur. volumetric solution
#: 31770.294
: VWR Chemicals
Vand, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
#: 83645.320
: VWR Chemicals