Pyronine G
Glass bottle for solids, 1 g
Katalog # CAYM14488-1
Leverandør: Cayman Chemical
Gå til:
Product Details & Documents
Synonymer: Pyronin Y, Pyronine Y, 3,6-Bis(dimethylamino)xanthylium chloride
- CAS nummer:92-32-0
- Formel:C₁₇H₁₉ClN₂O
- MDL nummer:MFCD00011725
- Molecular Weight:302.8 g/mol
- Merck Index:12,08190
- Smeltende Pt:250…260 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:202-147-8
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:1 g



