Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Polymyxin Bsulfat

Polymyxin Bsulfat

Glass bottle for solids, 1 g
Katalog # CAYM14157-1
Leverandør:   Cayman Chemical
Polymyxin Bsulfat
Katalog # CAYM14157-1
Leverandør:  Cayman Chemical
CAS nummer:  1405-20-5
Polymyxin Bsulfat

Glass bottle for solids, 1 g

CAS nummer:  1405-20-5
Price
687,00 kr
Log ind for tilgængelighed
1 * 1 g
Product Details & Documents
  • CAS nummer:1405-20-5
  • Formel:C₅₅H₉₆N₁₆O₁₃·2H₂SO₄
  • MDL nummer:MFCD00131991
  • Molecular Weight:1385.61 g/mol
  • Opbevaringstemperatur:Stue temperatur
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.