Gemfibrozil
Glass bottle for solids, 5 g
CAYM14835-5
: CI-719
- :25812-30-0
- :C₁₅H₂₂O₃
- :MFCD00079335
- :250.34 g/mol
- :12,04394
- :395 °C (1013 hPa)
- :59…61 °C
- :247-280-2
- Pakning:Glass bottle for solids
- Pakn:5 g