Gemfibrozil
Glass bottle for solids, 1 g
Katalog # CAYM14835-1
Leverandør: Cayman Chemical
Gå til:
Product Details & Documents
Synonymer: CI-719
- CAS nummer:25812-30-0
- Formel:C₁₅H₂₂O₃
- MDL nummer:MFCD00079335
- Molecular Weight:250.34 g/mol
- Merck Index:12,04394
- Kogende Pt:395 °C (1013 hPa)
- Smeltende Pt:59…61 °C
- EINECS:247-280-2
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:1 g



