4'-Hydroxy-3'-methoxyacetophenon
Glass bottle for solids, 10 g
Katalog # CAYM11976-10
Leverandør: Cayman Chemical
4'-Hydroxy-3'-methoxyacetophenon
Katalog # CAYM11976-10
Leverandør: Cayman Chemical
CAS nummer: 498-02-2
Gå til:
Product Details & Documents
Synonymer: 4-Hydroxy-3-methoxy-acetophenone, 1-(4-Hydroxy-3-methoxyphenyl)ethanone, Acetovanillone, Apocynin
- CAS nummer:498-02-2
- Formel:C₉H₁₀O₃
- MDL nummer:MFCD00008747
- Molecular Weight:166.18 g/mol
- Merck Index:13,00746
- Kogende Pt:295 °C (1013 hPa)
- Smeltende Pt:115 °C
- EINECS:207-854-5
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:10 g



