Orders LinkContactUsLinkComponent
Gemfibrozil
Glass bottle for solids, 25 g
  CAYM14835-25
Gemfibrozil
  CAYM14835-25
 :  25812-30-0
Gemfibrozil
Glass bottle for solids, 25 g
 :  25812-30-0
2.300,00 kr
1 * 25 g
 :  CI-719
  •  :25812-30-0
  •  :C₁₅H₂₂O₃
  •  :MFCD00079335
  •  :250.34 g/mol
  •  :12,04394
  •  :395 °C (1013 hPa)
  •  :59…61 °C
  •  :247-280-2
  • Pakning:
    Glass bottle for solids
  • Pakn:
    25 g