Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
2,4-Dihydroxy-5-fluorpyrimidin

2,4-Dihydroxy-5-fluorpyrimidin

Glass bottle for solids, 5 g
Katalog # CAYM14416-5
Giftig
Sundhed
Leverandør:   Cayman Chemical
2,4-Dihydroxy-5-fluorpyrimidin
Katalog # CAYM14416-5
Leverandør:  Cayman Chemical
CAS nummer:  51-21-8
2,4-Dihydroxy-5-fluorpyrimidin

Glass bottle for solids, 5 g

CAS nummer:  51-21-8
Price
592,00 kr
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents
Synonymer:  Fluorouracil, 5-Fluoropyrimidine-2,4(1H,3H)-dione, 5-FU
  • CAS nummer:51-21-8
  • Formel:C₄H₃FN₂O₂
  • MDL nummer:MFCD00006018
  • Molecular Weight:130.08 g/mol
  • Merck Index:13,04208
  • Kogende Pt:347 °C (1013 hPa)
  • Smeltende Pt:282…283 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-085-6
  • FN:2811
  • ADR:6.1,III
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    5 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.