Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
2,4-Dihydroxy-5-fluorpyrimidin

2,4-Dihydroxy-5-fluorpyrimidin

Glass bottle for solids, 10 g
Catalog # CAYM14416-10
Toxic
Health
Supplier:   Cayman Chemical
2,4-Dihydroxy-5-fluorpyrimidin
Catalog # CAYM14416-10
Supplier:  Cayman Chemical
CAS Number:  51-21-8
2,4-Dihydroxy-5-fluorpyrimidin

Glass bottle for solids, 10 g

CAS Number:  51-21-8
Price
1.140,00 kr
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  Fluorouracil, 5-Fluoropyrimidine-2,4(1H,3H)-dione, 5-FU
  • CAS Number:51-21-8
  • Formula:C₄H₃FN₂O₂
  • MDL Number:MFCD00006018
  • Molecular Weight:130.08 g/mol
  • Merck Index:13,04208
  • Boiling Pt:347 °C (1013 hPa)
  • Melting Pt:282…283 °C
  • Storage Temperature:Stue temperatur
  • EINECS:200-085-6
  • UN:2811
  • ADR:6.1,III
Specifications

Specifications

  • Pakning:
    Glass bottle for solids
  • Pakn:
    10 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.