Polymyxin Bsulfat
Glass bottle for solids, 500 mg
Katalog # CAYM14157-500
Leverandør: Cayman Chemical
Gå til:
Product Details & Documents
- CAS nummer:1405-20-5
- Formel:C₅₅H₉₆N₁₆O₁₃·2H₂SO₄
- MDL nummer:MFCD00131991
- Molecular Weight:1385.61 g/mol
- Opbevaringstemperatur:Stue temperatur
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:500 mg
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



