Orders LinkContactUsLinkComponent
Stoppers for sample bottles, BEHROTEST®
Stoppers for sample bottles, BEHROTEST®
24/29
Katalog # 719-1311
Leverandør:   BEHR
Stoppers for sample bottles, BEHROTEST®
Katalog # 719-1311
Leverandør:  BEHR
Stoppers for sample bottles, BEHROTEST®
24/29
Price
133,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These glass stoppers can be used instead of DIN/DEV diagonally cut stoppers. They are easy to use due to the lack of the easily broken edges.
  • No formation of bubbles below the stopper tip on inserting the stopper into the liquid sample
  • Less breakage or cracking risk than hollow stoppers
Specifikationer
  • NS:
    24/29