Ladies sweat jacket, Acode® Basecamp
mørkeblå, M
Katalog # KWIN110170543M
Leverandør: FRISTADS KANSAS
Gå til:
Product Details & Documents
Double-face sweat jacket made from 80% cotton and 20% polyester.
- Red contrast colour at collar and zip pullers
- Two front pockets, one breast pocket with zip
- Cuffs and hem in rib with elastane
- Tapered waist
Weight: 320 g/m²
Documents
Specifikationer
- Tøj størrelse:M
- Farve:mørkeblå
- Seq:02
- Køn:For kvinder
- Cat. No.:KWIN110170543M
- Sleeve or leg length:Long
- Vægt:320 g/m²
- Outfit applikationer:General Purpose
- Lukkestørrelse:Med lynlås
- Stof:80% cotton and 20% polyester