Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Scrapers, SteriWare®

Non sterile, 225 mm, 110×110 mm
Katalog # SAMP8785H-110
Leverandør:   Sampling Systems
Scrapers, SteriWare®
Katalog # SAMP8785H-110
Leverandør:  Sampling Systems
Scrapers, SteriWare®
Scrapers, SteriWare®

Non sterile, 225 mm, 110×110 mm

Price
2.620,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents

These disposable scrapers are made from high quality HDPE. They are ideal in the manufacture of creams, ointments and pastes.

  • Moulded & packed in a cleanroom
  • Made from FDA and EU conforming HDPE
  • BSE/TSE-free
  • Available as gamma irradiated sterile (SAL 10-6)
  • Cross-contamination risk is eliminated - the device can be used straight from the bag
  • No cleaning is required before or after sampling

Full batch traceability

 : Individually bagged.

Specifikationer
  • Aftørringsområde ca.:
    110×110 mm
  • Beskrivelse:
    Non sterile
  • Længde:
    225 mm
  • Seq:
    03
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.