Orders LinkContactUsLinkComponent
Basic Laboratory Chairs
Chair, with contact backrest, 470 - 610 mm
Katalog # BIMO9134-2571
Leverandør:   bimos
Basic Laboratory Chairs
Katalog # BIMO9134-2571
Leverandør:  bimos
Basic Laboratory Chairs
Basic Laboratory Chairs
Chair, with contact backrest, 470 - 610 mm
Price
4.570,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
The tried-and-tested all-rounder for use in laboratories is a great entry level laboratory chair, perfect for a range of workplace environments.
  • With a sturdy steel 5 star base
  • Available in two different frame colours: black or light grey
  • Choice of three different mechanisms: contact backrest, contact backrest with seat tilt adjustment and synchronous mechanism with weight regulation
Basic Laboratory can be supplied with Stamskin synthetic leather or with fabric upholstery. Not only is Stamskin Top easy to clean, but it also has antibacterial, antifungal and antimicrobial properties.
Conform to: DIN 68 877, German GS „safety tested“ certificate and Quality Management System DIN EN ISO 9001
Certifications: Conform to: DIN 68 877, German GS 'safety tested' certificate and Quality Management System DIN EN ISO 9001
Specifikationer
  • Beskrivelse:
    Chair, with contact backrest
  • Højde:
    470 - 610 mm
  • Seq:
    02