Orders LinkContactUsLinkComponent
Connection system GL 45
Bottle pourer, PTFE, GL 45, with venting tube and attached PP blue cap
Katalog # BIBB4542/02
Leverandør:   SciLabware
Connection system GL 45
Katalog # BIBB4542/02
Leverandør:  SciLabware
Connection system GL 45
Connection system GL 45
Bottle pourer, PTFE, GL 45, with venting tube and attached PP blue cap
Price
1.380,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
PP screwcaps for Pyrex® GL 45 media-lab bottles facilitate the transfer of liquids within a closed and sterile system.
  • Temperature resistant to 140 °C
  • Membrane screw cap is ideal for aerobic microbiology and cell culture, with gas permeable
  • Dishwasher safe
Ideal for chemistry and biotechnology laboratories. Components are fully interchangeable and compatible with both two and three port screwcaps. Four different tubing diameters can be accommodated (Ext.Ø; 1,6, 3,0, 3,2 and 6,0 mm). The additional pressure compensation set that includes a membrane filter with 0,2 μm sized pores permits sterile pressure equalisation. Unused ports can be sealed with a blanking cap. Components are fully interchangeable and compatible with both two and three port screwcaps.
Specifikationer
  • Beskrivelse:
    Bottle pourer, PTFE, GL 45, with venting tube and attached PP blue cap
  • Seq:
    14