Venepunktursæt, Surshield™ Surflo®
Winged needles with protector, G 19, hvid, 19 mm
Katalog # TERUSV-S19NL30
Leverandør: TERUMO
Gå til:
Product Details & Documents
Surshield prevents needle stick injuries with painless placement.
Cannula sharpness combined with a special silicone coating allows an easy and smooth placement.
Specifikationer
- Måler:G 19
- Farve:hvid
- Beskrivelse:Winged needles with protector
- Længde:19 mm
- Seq:001