Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Drenchers

Drencher, straight, 25 mm, 0,9 mm
  SOCO376.0925S
Drenchers
  SOCO376.0925S
Drenchers
Drenchers

Drencher, straight, 25 mm, 0,9 mm

 
225,00 kr
   
1 * 3 Items
 

These straight or bent drenchers are made from stainless steel. They are ideal for safe animal nose and mouth feeding.

  • Luer lock fittings
 
  • Udv.Ø:
    0,9 mm
  • Beskrivelse:
    Drencher, straight
  • Længde:
    25 mm
  • Seq:
    05
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.