Reagent containers for Microlab® STAR line
Reagent container without lid, self-standing, 1T, Self-standing, 1T, 60 ml, 89,7×20,0×64,8 mm, transparent, Individually wrapped
Leverandør: HAMILTON ROBOTICS
Reagent containers for Microlab® STAR line
Katalog # 736-0830
Leverandør: HAMILTON ROBOTICS
Marketsource
Gå til:
Product Details & Documents
PP, white or black. These reagent reservoirs with 60, 120 or 200 ml capacity can be placed in a reagent carrier on the STAR line instruments. The reservoirs are available self-standing or non-self-standing.
- Special bottom geometry minimises residual volumes
- With volume markings, without or with lid
- With one (60 ml), three (120 ml) or four (200 ml) wave breakers to avoid splashes during movements of the filled containers
- IVD compliant
- Autoclavable
Bestillingsoplysninger: The carriers are not included and need to be ordered separately.
Documents
Specifikationer
- Pakket:Individually wrapped
- Model:Self-standing, 1T
- Kapacitet:60 ml
- Farve:transparent
- Beskrivelse:Reagent container without lid, self-standing, 1T
- Seq:1
- B×D×H:89,7×20,0×64,8 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



