Reagent containers for Microlab® STAR line
Reagent container with lid, self-standing, 1T, Self-standing, 1T, with lid, 60 ml, 89,7×20,0×64,8 mm, gennemsigtig, Individually wrapped
Katalog # 736-0831
Marketsource
Leverandør: HAMILTON ROBOTICS
Reagent containers for Microlab® STAR line
Katalog # 736-0831
Leverandør: HAMILTON ROBOTICS
Marketsource
Gå til:
Product Details & Documents
PP, white or black. These reagent reservoirs with 60, 120 or 200 ml capacity can be placed in a reagent carrier on the STAR line instruments. The reservoirs are available self-standing or non-self-standing.
- Special bottom geometry minimises residual volumes
- With volume markings, without or with lid
- With one (60 ml), three (120 ml) or four (200 ml) wave breakers to avoid splashes during movements of the filled containers
- IVD compliant
- Autoclavable
: The carriers are not included and need to be ordered separately.
Specifikationer
- Pakket:Individually wrapped
- Model:Self-standing, 1T, with lid
- Kapacitet:60 ml
- Farve:gennemsigtig
- Beskrivelse:Reagent container with lid, self-standing, 1T
- Seq:2
- B×D×H:89,7×20,0×64,8 mm



