Test charts
Test charts with optical brightner, sort/hvid ternet, A3, 297×420 mm
SIMEVF2354
Test charts are used for testing physical properties of coating, lacquers, and inks.
- Dimensions from A6 up to A4
- Film laminated for an excellent solvent, chemical resistance, and film spread
- Section for the date, time, and test number
Suitable for determining hiding power, opacity, and spreading rate in coating laboratories and paint production.
: A certificate is available for charts with an optical brightener.
- Format:A3
- Farve:sort/hvid ternet
- Beskrivelse:Test charts with optical brightner
- Seq:11
- B×H:297×420 mm